MAN372905626 Manchester Organics

Chloromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether

  • MDL number MFCD09952406
  • Cas Number : 26103-07-1
  • Molecular Weight : 216.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/S26275
Availability details for
Quantity
  • Availability
  • Add to Cart