MAN372905736 Manchester Organics

4-Fluorocyclohexanone

  • MDL number MFCD11520472
  • Cas Number : 68223-64-3
  • Molecular Weight : 116.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/T15530
Availability details for
Quantity
  • Availability
  • Add to Cart