MAN372905951 Manchester Organics

4,6-Dichloro-3-methylpyridazine

  • MDL number MFCD21604745
  • Cas Number : 68240-43-7
  • Molecular Weight : 163.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y10519
Availability details for
Quantity
  • Availability
  • Add to Cart