MAN372905989 Manchester Organics

3-Methyl-5-(tributylstannyl)isoxazole

  • MDL number MFCD01319023
  • Molecular Weight : 372.133

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.85
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B26733
Availability details for
Quantity
  • Availability
  • Add to Cart