MAN372906048 Manchester Organics

(5S)-5-(Trifluoromethyl)pyrrolidin-2-one

  • MDL number MFCD22566326
  • Cas Number : 1287211-10-2
  • Molecular Weight : 153.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 9798
Country of Origin GB
Product Link https://www.manchesterorganics.com/W15533
Availability details for
Quantity
  • Availability
  • Add to Cart