MAN372906098 Manchester Organics

5,5,6,6,6-Pentafluorohexane-2,4-dione

  • MDL number MFCD00466808
  • Cas Number : 356-40-1
  • Molecular Weight : 204.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/C27089
Availability details for
Quantity
  • Availability
  • Add to Cart