MAN372906141 Manchester Organics

2-Amino-N-methyl-acetamide hydrochloride

  • MDL number MFCD00153440
  • Molecular Weight : 124.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J23593
Availability details for
Quantity
  • Availability
  • Add to Cart