MAN372906220 Manchester Organics

4-(Perfluorocyclohexyl)butan-1-ol

  • MDL number MFCD06248652
  • Molecular Weight : 354.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X27132
Availability details for
Quantity
  • Availability
  • Add to Cart