MAN372906398 Manchester Organics

9-Bromonon-1-ene

  • MDL number MFCD09037826
  • Cas Number : 89359-54-6
  • Molecular Weight : 205.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/G31057
Availability details for
Quantity
  • Availability
  • Add to Cart