MAN372906528 Manchester Organics

3-[2-(Trifluoromethyl)phenyl]oxetan-3-amine hydrochloride

  • MDL number MFCD20233042
  • Cas Number : 1349715-87-2
  • Molecular Weight : 253.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J27308
Availability details for
Quantity
  • Availability
  • Add to Cart