MAN372906785 Manchester Organics

1-(2,6-Difluorophenyl)ethanol

  • MDL number MFCD00274321
  • Cas Number : 87327-65-9
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/B31305
Availability details for
Quantity
  • Availability
  • Add to Cart