MAN372906857 Manchester Organics

2,4-Dichloro-5-fluorobenzoic acid

  • MDL number MFCD00075281
  • Molecular Weight : 209.00192

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B31374

Safety Information

MSDS https://www.manchesterorganics.com/B31374
Availability details for
Quantity
  • Availability
  • Add to Cart