MAN372906863 Manchester Organics

2-Methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide

  • MDL number MFCD14702522
  • Molecular Weight : 175.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L13916
Availability details for
Quantity
  • Availability
  • Add to Cart