MAN372906880 Manchester Organics

2-(Oxetan-3-yl)acetic acid

  • MDL number MFCD19442256
  • Molecular Weight : 116.11522

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V13924

Safety Information

MSDS https://www.manchesterorganics.com/V13924
Availability details for
Quantity
  • Availability
  • Add to Cart