MAN372906923 Manchester Organics

(3-Methyloxetan-3-yl)methanamine

  • MDL number MFCD08703640
  • Molecular Weight : 101.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z13928
Availability details for
Quantity
  • Availability
  • Add to Cart