MAN372906924 Manchester Organics

3-Methyloxetan-3-ol

  • MDL number MFCD18909291
  • Cas Number : 162816-08-2
  • Molecular Weight : 88.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/B13930
Availability details for
Quantity
  • Availability
  • Add to Cart