MAN372906943 Manchester Organics

3-(Aminomethyl)oxetan-3-ol

  • MDL number MFCD20444451
  • Cas Number : 1305208-47-2
  • Molecular Weight : 103.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/K13938
Availability details for
Quantity
  • Availability
  • Add to Cart