MAN372907173 Manchester Organics

3-Methoxypyrrolidine

  • MDL number MFCD07787006
  • Molecular Weight : 101.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D31629
Availability details for
Quantity
  • Availability
  • Add to Cart