MAN372907269 Manchester Organics

4-Nitro-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD07778365
  • Molecular Weight : 163.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W31737
Availability details for
Quantity
  • Availability
  • Add to Cart