MAN372907306 Manchester Organics

[(3R)-Pyrrolidin-3-yl]methanol hydrochloride

  • MDL number MFCD09800470
  • Molecular Weight : 137.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/L31774
Availability details for
Quantity
  • Availability
  • Add to Cart