MAN372907335 Manchester Organics

(2R,4S)-4-Hydroxypyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl

  • MDL number MFCD09954932
  • Cas Number : 135042-17-0
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/T31803
Availability details for
Quantity
  • Availability
  • Add to Cart