MAN375326011 Manchester Organics

4-Amino-3-hydroxypiperidine

  • MDL number MFCD22533520
  • Cas Number : 1391004-19-5
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-aminopiperidin-3-ol
Inchi Code 1S/C5H12N2O/c6-4-1-2-7-3-5(4)8/h4-5,7-8H,1-3,6H2
Inchi Key UVBKPWNFXFUDOU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/F12230
Availability details for
Quantity
  • Availability
  • Add to Cart