MAN375326118 Manchester Organics

Cyclobutanecarbonitrile

  • MDL number MFCD00013735
  • Molecular Weight : 81.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W26538
Availability details for
Quantity
  • Availability
  • Add to Cart