MAN375326209 Manchester Organics

N-Methylmethanesulfonamide

  • MDL number MFCD00776277
  • Molecular Weight : 109.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X32014
Availability details for
Quantity
  • Availability
  • Add to Cart