MAN375326320 Manchester Organics

2,3,5-Trimethyl-1,4-benzoquinone

  • MDL number MFCD00045537
  • Molecular Weight : 150.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N32121
Availability details for
Quantity
  • Availability
  • Add to Cart