MAN375326394 Manchester Organics

3,5-Difluoro-4-methoxy-benzaldehyde

  • MDL number MFCD04115911
  • Molecular Weight : 172.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U32195
Availability details for
Quantity
  • Availability
  • Add to Cart