MAN375326415 Manchester Organics

2,4-Dichloro-5-iodopyridine

  • MDL number MFCD09033755
  • Cas Number : 343781-49-7
  • Molecular Weight : 273.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/E12229
Availability details for
Quantity
  • Availability
  • Add to Cart