MAN375326525 Manchester Organics

N1,N2-Dimethylcyclohexane-1,2-diamine

  • MDL number MFCD05664382
  • Molecular Weight : 142.242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F32298
Availability details for
Quantity
  • Availability
  • Add to Cart