MAN375326593 Manchester Organics

5-(Diethylamino)pentylamine

  • MDL number MFCD00047979
  • Cas Number : 34987-15-0
  • Molecular Weight : 158.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N~1~,N~1~-diethyl-1,5-pentanediamine
Inchi Code 1S/C9H22N2/c1-3-11(4-2)9-7-5-6-8-10/h3-10H2,1-2H3
Inchi Key JYCCKJSLUHEIDU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/H12232
Availability details for
Quantity
  • Availability
  • Add to Cart