MAN375326647 Manchester Organics

(R)-tert-Butyl 2-phenylpiperazine-1-carboxylate

  • MDL number MFCD04972241
  • Molecular Weight : 262.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D27328
Availability details for
Quantity
  • Availability
  • Add to Cart