MAN375326654 Manchester Organics

4-(Pentafluoroethyl)benzylamine

  • MDL number MFCD22416533
  • Molecular Weight : 225.15853

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T15555

Safety Information

MSDS https://www.manchesterorganics.com/T15555
Availability details for
Quantity
  • Availability
  • Add to Cart