MAN375326657 Manchester Organics

(2Z)-3-(Diethylamino)-2-fluoroprop-2-enal

  • MDL number MFCD18633001
  • Molecular Weight : 145.175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V15557
Availability details for
Quantity
  • Availability
  • Add to Cart