MAN375326694 Manchester Organics

1-(Difluoromethoxy)-3-fluoro-2-methylbenzene

  • MDL number MFCD14698458
  • Cas Number : 1214333-52-4
  • Molecular Weight : 176.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name difluoromethyl 3-fluoro-2-methylphenyl ether
Inchi Code 1S/C8H7F3O/c1-5-6(9)3-2-4-7(5)12-8(10)11/h2-4,8H,1H3
Inchi Key SJLJZJPOEVOMRT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/B15586
Availability details for
Quantity
  • Availability
  • Add to Cart