MAN375326786 Manchester Organics

1,2-Phenylenediacetonitrile

  • MDL number MFCD00001905
  • Molecular Weight : 156.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin GB
Product Link https://www.manchesterorganics.com/R32376
Availability details for
Quantity
  • Availability
  • Add to Cart