MAN375326787 Manchester Organics

4-(Diethylamino)butan-1-ol

  • MDL number MFCD00053355
  • Cas Number : 2683-56-9
  • Molecular Weight : 145.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(diethylamino)-1-butanol
Inchi Code 1S/C8H19NO/c1-3-9(4-2)7-5-6-8-10/h10H,3-8H2,1-2H3
Inchi Key KHYKXDSWXWVQTA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/J12233
Availability details for
Quantity
  • Availability
  • Add to Cart