MAN375326846 Manchester Organics

(6-Hydroxy-2-naphthyl)boronic acid

  • MDL number MFCD00092986
  • Cas Number : 173194-95-1
  • Molecular Weight : 187.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/G32391
Availability details for
Quantity
  • Availability
  • Add to Cart