MAN375326861 Manchester Organics

(5-Isopropoxy-3-pyridyl)boronic acid

  • MDL number MFCD15529675
  • Cas Number : 850991-41-2
  • Molecular Weight : 181

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-isopropoxypyridin-3-ylboronic acid
Inchi Code 1S/C8H12BNO3/c1-6(2)13-8-3-7(9(11)12)4-10-5-8/h3-6,11-12H,1-2H3
Inchi Key XQHUPBAXLPBXOT-UHFFFAOYSA-N

Description

Purity 94%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y32406
Availability details for
Quantity
  • Availability
  • Add to Cart