MAN375326878 Manchester Organics

(5-Bromo-2-methyl-phenyl)boronic acid

  • MDL number MFCD16295061
  • Cas Number : 774608-13-8
  • Molecular Weight : 214.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2-methylphenylboronic acid
Inchi Code 1S/C7H8BBrO2/c1-5-2-3-6(9)4-7(5)8(10)11/h2-4,10-11H,1H3
Inchi Key QOZFXELTNKMOQP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/S32423
Availability details for
Quantity
  • Availability
  • Add to Cart