MAN375327000 Manchester Organics

(4-Benzyloxy-3,5-difluoro-phenyl)boronic acid

  • MDL number MFCD16295234
  • Cas Number : 156635-88-0
  • Molecular Weight : 264.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(benzyloxy)-3,5-difluorophenylboronic acid
Inchi Code 1S/C13H11BF2O3/c15-11-6-10(14(17)18)7-12(16)13(11)19-8-9-4-2-1-3-5-9/h1-7,17-18H,8H2
Inchi Key YWUHRGQYLQMLHW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z32545
Availability details for
Quantity
  • Availability
  • Add to Cart