MAN375327399 Manchester Organics

1-Bromo-3-fluoro-5-(trifluoromethoxy)benzene

  • MDL number MFCD22551456
  • Cas Number : 1129541-09-8
  • Molecular Weight : 259

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D15872
Availability details for
Quantity
  • Availability
  • Add to Cart