MAN375327499 Manchester Organics

(2-Cyano-3-pyridyl)boronic acid

  • MDL number MFCD08458475
  • Cas Number : 874290-88-7
  • Molecular Weight : 147.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N32811
Availability details for
Quantity
  • Availability
  • Add to Cart