MAN375327537 Manchester Organics

2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

  • MDL number MFCD11856048
  • Cas Number : 1003845-08-6
  • Molecular Weight : 240.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/W32841
Availability details for
Quantity
  • Availability
  • Add to Cart