MAN375327585 Manchester Organics

(2-Chloro-4-hydroxy-phenyl)boronic acid

  • MDL number MFCD02684320
  • Cas Number : 766549-26-2
  • Molecular Weight : 172.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/P32881
Availability details for
Quantity
  • Availability
  • Add to Cart