MAN375327640 Manchester Organics

(2,5-Dibromophenyl)boronic acid

  • MDL number MFCD09743927
  • Cas Number : 1008106-93-1
  • Molecular Weight : 279.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-dibromophenylboronic acid
Inchi Code 1S/C6H5BBr2O2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,10-11H
Inchi Key CMLBXUUGERIZOO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z32936
Availability details for
Quantity
  • Availability
  • Add to Cart