MAN375327728 Manchester Organics

(2-Aminopyridin-5-yl)boronic acid

  • MDL number MFCD09907980
  • Cas Number : 851524-96-4
  • Molecular Weight : 137.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/E26594
Availability details for
Quantity
  • Availability
  • Add to Cart