MAN375327787 Manchester Organics

4-Borono-2-nitro-benzoic acid

  • MDL number MFCD10696661
  • Cas Number : 80500-28-3
  • Molecular Weight : 210.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/J32623
Availability details for
Quantity
  • Availability
  • Add to Cart