MAN375327858 Manchester Organics

1-Bromo-4-[chloro(difluoro)methoxy]benzene

  • MDL number MFCD22580792
  • Cas Number : 112556-13-5
  • Molecular Weight : 257.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D15801
Availability details for
Quantity
  • Availability
  • Add to Cart