MAN375327941 Manchester Organics

(3-Chloro-2-formyl-phenyl)boronic acid

  • MDL number MFCD13181639
  • Molecular Weight : 184.385

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C32709
Availability details for
Quantity
  • Availability
  • Add to Cart