MAN375328065 Manchester Organics

1-Bromo-3-chloro-5-(trifluoromethoxy)benzene

  • MDL number MFCD11856023
  • Cas Number : 1417567-41-9
  • Molecular Weight : 275.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N15952
Availability details for
Quantity
  • Availability
  • Add to Cart