MAN375328149 Manchester Organics

6-Methoxynaphthalene-2-sulfonyl chloride

  • MDL number MFCD12402643
  • Cas Number : 56875-59-3
  • Molecular Weight : 256.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methoxy-2-naphthalenesulfonyl chloride
Inchi Code 1S/C11H9ClO3S/c1-15-10-4-2-9-7-11(16(12,13)14)5-3-8(9)6-10/h2-7H,1H3
Inchi Key QYCDJXYAIDWYFK-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/Z33097
Availability details for
Quantity
  • Availability
  • Add to Cart